Learn More
ATCC Escherichia coli; JM107
Supplier: ATCC 47014
Escherichia coli strain JM107 is a bacterial strain that can be used as a transformation host in molecular biology applications or as a host for M13 and other filamentous bacteriophages.
This item is not returnable.
View return policy