Learn More
Hach Company Standard Cell Solution, Concentrated pH 7.0 Buffer (Equi-Transferrant), 500 mL
Supplier:  Hach Company 25M1A1025115
Encompass_Preferred
Standard Cell Solution for Hach pHD Differential pH and ORP Sensors, packaged in resealable 500 mL bottle. Used to replenish standard cell chamber while replacing salt bridge.
Catalog No. 50-199-7405
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.