Learn More
Enzo Life Sciences EQ0 NHS ester (1mg)
SDP
Supplier:  Enzo Life Sciences ENZCHM2080001
Encompass_Preferred
EQ0 NHS ester is an amine-reactive quencher dye. It has a broad visible absorption but no detectable fluorescence emission, making it useful as an acceptor in fluorescence  resonance energy transfer (FRET) applications. It can be used to quench reporter dyes: AMCA, AMCA-X, DEAC, DEAC-X, MCA, MCC, CF™ 350, CF™405S, CF™405M, Pacific Blue™, Cascade Blue, Marina Blue, Alexa Fluor 350, Alexa Fluor 405, DyLight 350, DyLight 405, ATTO 390, ATTO 425, ATTO 465, iFluor™350, iFLuor™405, BD Horizon V450, BUV395, eFluor450. Purity: ≥80% (HPLC). Solubility: Soluble in DMSO or DMF. Long Term Storage: -20°C. Applications: Fluorescent detection FRET.
Catalog No. 50-201-3533
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.