Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Eppendorf Tubes™, 5.0 mL
5.0 mL Eppendorf Tubes™ enable the simple and safe processing of sample volumes up to 5.0 mL with a broad operating temperature range, centrifugation safety and excellent sample recovery.
$101.80 - $133.00
Specifications
Capacity (Metric) | 5.0 mL |
---|---|
Color | Clear |
Material | Polypropylene |
For Use With (Application) | Easy processing of samples up to 5.0 mL, DNA isolation, especially with samples of limited quantity of source material, Preparation of mastermixes and buffers, Cell and tissue lysis |
For Use With (Equipment) | Centrifuge |
Catalog Number | Mfr. No. | Clean Claims | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Clean Claims | Price | Quantity & Availability | |||||
13-864-405
|
Eppendorf™
0030122305 |
Eppendorf Quality™ |
Each for $101.80
|
|
|||||
13-864-406
|
Eppendorf™
0030122313 |
PCR Clean |
Each for $125.30
|
|
|||||
13-864-407
|
Eppendorf™
0030122321 |
Pyrogen, Dnase, Rnase, human and bacterial DNA free |
Each for $133.00
|
|
|||||
Description
Features:
- Ergonomic operation:
- Snap Cap - Simple, practical and ergonomic single-handed operation.
- Screw Cap - Flattened and grooved sides provide optimal grip and user-friendly handling.
- Broad operating temperature range:
- Screw Cap Tubes: −86 to 100°C
- Snap Cap Tubes: −86 to 80°C (When using the clips ≤100°C)
- Material: High-quality, transparent polypropylene, free of plasticizers, biocides or mold release agents, for reliable test results.
- Certified purity: Available in lot specific certified Eppendorf Quality™, PCR Clean, Sterile, and Eppendorf Biopur™ purity grades.
- Centrifugation safety: Safety and stability for centrifugation up to 25,000 × g. Fast and efficient protocols.
- Conical shape: Compatible with accessories for conical 15 mL tubes - many existing adapters and racks can be used.
- Sample recovery: Available in Eppendorf LoBind™ material for maximum recovery of valuable samples.
Applications:
- Easy processing of samples up to 5.0 mL
- High yields in DNA isolation, especially with samples with limited quantity of source material
- Convenient and safe cell culture applications
- Easy sample access for low risk of contamination
- Preparation of mastermixes and buffers
- Safe cell and tissue lysis
Specifications
5.0 mL | |
Polypropylene | |
Centrifuge | |
Bags | |
Yes | |
Conical |
Clear | |
Easy processing of samples up to 5.0 mL, DNA isolation, especially with samples of limited quantity of source material, Preparation of mastermixes and buffers, Cell and tissue lysis | |
200 Tubes (2 Bags x 100 Tubes) | |
−86°C to +100°C | |
Screw Cap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title