Learn More
LGC Biosearch Technologies CopyCutter EPI400 Chemically Competent E. coli, 10 Rxns
Supplier:  LGC Biosearch Technologies C400CH10
Encompass_Preferred
CopyCutter™ EPI400™ E. coli cells will significantly lower the copy number of a wide variety of common vectors to enable cloning of unstable DNA sequences and those expressing toxic proteins
Catalog No. NC9224981
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.