Learn More
Electron Microscopy Sciences Enviro-Genie 6-in-1, 120 V
Supplier: Electron Microscopy Sciences 7183510
Designed for precise temperature control and uniformity for demanding incubation/refrigeration application. Uses Thermo-Electric (Peltier) refrigeration and heating without a compressor. Chamber air is gently and continuously recirculated to ensure temperature consistency and uniformity. Re-circulated air eliminates the possibility of contamination from outside air. Shatter resistant viewing window allows for inspection of samples without effecting the chamber environment.
Applications include the study of microorganisms and tissue cell growth under strictly controlled conditions of temperature and agitation. It performs membrane based assays/washing and hybridizations more efficiently. Microprocessor controls easily achieve desired rotating, rocking, stirring, shaking, temperature and time; all of which are displayed on large LED's for accurate reproducible results.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.