Learn More
New England Biolabs, Inc. EnGen SpRY Cas9 - 500 pmol
SDP
Supplier:  New England Biolabs, Inc. M0669T
Encompass
EnGen SpRY Cas9 from Streptococcus pyogenes is an engineered, RNA-guided, DNA endonuclease that catalyzes site-specific cleavage of double-stranded DNA (dsDNA).
Catalog No. 50-258-1809
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.