Learn More
Terumo Engauge Safety Hypodermic Syringe with Safety Needle
Engauge Safety Hypodermic Syringe with Safety Needle features three different modes of activation: Thumb, Finger and Hard Surface.
Supplier:  Terumo SENG01L2525R
 View more versions of this product
Catalog No. 22-290-030
This item is not returnable. View return policy
Description
Description
  • Low Dead Space
  • 3 modes of activation: Thumb, Finger and Hard Surface
  • Economical option
Specifications
Specifications
Not Autoclavable
25 G
1 in.
1 mL
Hypodermic Needle
Yes
Polypropylene (Barrel), Polypropylene (Plunger), Polypropylene (Hub)
Yes
Single-use
Bevel
Luer Lock
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.