Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Buffered Neutral Formalin (10%)

Buffered Neutral Formalin (10%)

$173.00 - $338.00

Specifications

Concentration 10%
Type Buffered Neutral Formalin (10%)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity & Availability  
ES9720
View Documents
ENG Scientific
9720
1 x 19 L (5 gal.) Cubetainer
Each for $338.00
missing translation for 'QuantityLeft'
Add to Cart
 
ES9700
View Documents
ENG Scientific
9700
2 x 950 mL Bottle
Pack of 2 for $173.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

  • Buffered Neutral Formalin (10%)
Specifications

Specifications

10%
Buffered Neutral Formalin (10%)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.