Learn More
Eurofins Discoverx Endothelin-1, 10 μg. (92-1164)
Supplier:  Eurofins Discoverx 921164
Encompass_Preferred
Endothelin 1 is an endothelium derived vasoconstrictive peptide produced in vascular endothelial cells. It is one of the three isoforms of endothelin, and a potent agonist of the endothelin receptors type A and type B (EDNRA/ETA and EDNRB/ETB). Widely expressed in all tissues, and responsible for maintenance of vascular tone. Inhibitors and antagonists are used to treat pulmonary hypertension. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9692
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.