Learn More
Electron Microscopy Sciences EMS Stainless steel Skin Punch
Supplier:  Electron Microscopy Sciences 69101
Encompass_Preferred
This skin punch will pierce dense skin with ease. The rotating hole template offers five hole sizes (1 mm, 1.2 mm, 1.4 mm, 1.7 mm, and 2 mm). Made of Stainless steel. OAL is 6.5" Long (165 mm).
Catalog No. 50-193-1268
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.