Learn More
MilliporeSigma™ Qualification Workbook
Provides instructions on performing system qual and completing qual forms
Supplier:  MilliporeSigma™ VRQSX005Y
 View more versions of this product
Catalog No. VRQSX005Y
Explore all special offers
Add to Cart
Includes: IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Description
Description
  • For use with Milli-RO, Milli-RX, Milli-Q, RiOs, SDS, Elix, Direct-Q systems
Specifications
Specifications
Provides instructions on performing system qual and completing qual forms
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Direct-Q 5 System
Qualification Workbook
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.