Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ pSTBlue-1 Perfectly Blunt Cloning Kit

$518.79 - $846.29

Specifications

Content And Storage Store at _70°C. Avoid freeze/thaw cycles.
For Use With (Application) Simplified cloning of DNA generated by PCR using any type of DNA polymerase
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-191-3
View Documents
MilliporeSigma™
701913
20 Reactions
Each for $518.79
Add to Cart
 
70-191-4
View Documents
MilliporeSigma™
701914
40 Reactions
Each for $846.29
Add to Cart
 
Description

Description

The pSTBlue-1 vector is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
Specifications

Specifications

Store at _70°C. Avoid freeze/thaw cycles.
Simplified cloning of DNA generated by PCR using any type of DNA polymerase
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.