Learn More
MilliporeSigma™ pSTBlue-1 AccepTor Linearized Vectors
$272.00 - $430.00
Description
The pSTBlue-1 is a general purpose vector with dual opposed T7 and SP6 promoters, both with amp and kan resistance, and an array of flanking restriction sites.For Research Use Only
Spot an opportunity for improvement?
Share a Content Correction