Learn More
Electron Microscopy Sciences Embedding Ring, Pink
Supplier: Electron Microscopy Sciences 62350P
Molded from the highest quality plastic, our Embedding Rings fit most base molds. The large writing surface further eases identification and handling. The base measures 1 5/8" x 1 1/8" and the ring measures 1" x 1" (25 x 25 mm).