Learn More
Biomatik Corporation Bovine Transforming Growth Factors Β2, Tgfβ2 Elisa Kit
Supplier:  Biomatik Corporation EKC31811
Encompass_Preferred
Bovine Transforming Growth Factors Β2, Tgfβ2 Elisa Kit for detecting Transforming Growth Factor, Beta 2 in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 6.25-100ng/ml. Sensitivity: 6.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P21214; TGFB2. Target Synonyms: TGFB2; Transforming growth factor beta-2 proprotein; Milk growth factor; MGF) [Cleaved into: Latency-associated peptide; LAP); Transforming growth factor beta-2; TGF-beta-2)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7230
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.