Learn More
Thermo Scientific HR-ICP-MS Instrument Torch | | 1 EA
Supplier: Thermo Scientific 1091250
Maintain consistently high performance by using only top quality dedicated consumables. The Guard Electrode GE feature decreases the ion energy spread to increase ion transmission for improved sensitivity. The GE feature is a requirement for Cold Plasma measurements.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.