Learn More
Electron Microscopy Sciences Ehrlich's Hematoxylin 1 LT
Supplier: Electron Microscopy Sciences 2604006
Ehrlich's Hematoxylin solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used to selectivity stain for nuclei.