Learn More
Hach Company EDTA Standard Solution, 0.0200N, 200 mL
Supplier: Hach Company 20529
For hardness determination by buret titration. Hach methods 8222 (calcium) and 8226 (total). 0.0200N (0.0100 M) (N/50) 200 mL bottle