Learn More
Across International ECO 150C 3.2 Cu Ft Vacuum Oven 3 Heated Shelves & LED Lights
Supplier: Across International AT32E
With Ai internal shelf heating technology, each shelf in these ovens has its own heater, in-shelf temperature sensor and temperature controller, together with great thermal-conductive aluminum and our new low proportional controller technologies, the result is perfect uniformity, accurate temperature, super-fast heating rates, minimum heat loss/exterior temperature and very low power consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.