Learn More
Bioanalytical Systems Inc EC U-ELECT-MPTE-10 UM DIA
SDP
Supplier:  Bioanalytical Systems Inc MF2005
Encompass_Preferred
Microelectrode; MPTE platinum; 10um diameter; For use with Low Current Module *Does not ship to Puerto Rico
Catalog No. 50-854-542
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.