Learn More
STEMCELL Technologies EasySep Mouse PE Positive Selection Kit II, For processing: 5 x 10 9 cells
Supplier: STEMCELL Technologies 17696
The EasySep Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.