Learn More
STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit, 60 mL buffy coat or whole blood
SDP
Supplier:  STEMCELL Technologies 17874
Encompass
The EasySep™ HLA Chimerism Whole Blood CD19 Positive Selection Kit is designed to isolate CD19+ cells from whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD19 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD19 antigen is present on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.This product replaces the EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084) for even faster cell isolations.
Catalog No. 50-197-4576
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.