Learn More
STEMCELL Technologies EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit, 2 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19232
Encompass
The EasySep™ Human CD4+CD127lowCD49d- Regulatory T Cell Enrichment Kit is designed to isolate CD4+CD127lowCD49d- cells from fresh peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells, CD127high, CD49d+ cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.
Catalog No. NC0594850
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.