Learn More
STEMCELL Technologies EasySep™ Human B Cell Enrichment Kit, For processing: 1 x 10^9 cells
Supplier: STEMCELL Technologies 19054
The EasySep™ Human B Cell Enrichment Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and dextran-coated magnetic particles. The labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human B Cell Isolation Kit (17954) (/products/easysep-human-b-cell-isolation-kit.html), which isolates cells in just 9 minutes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.