Learn More
Electron Microscopy Sciences Easy-Molds™ for size 00 Capsules, 8 mm diameter
Supplier:  Electron Microscopy Sciences 6993125
Encompass_Preferred
A newly designed mold which is convenient and offers better embedments. This tray-and-mold in one unit offers a unique specimen block that no other mold can. Each tray contains 2x10 block molds and measures 87x66 mm. Size 8mm diameter corresponds to capsule size "00" and forms a block face of 3mm.
Catalog No. 50-257-03
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.