Learn More
New England Biolabs, Inc. NEBExpress E. coli Lysis Reagent 100 mL
SDP
Supplier:  New England Biolabs, Inc. P8116S
Encompass
NEBExpress E. coli Lysis Reagent is a chemical lysis solution composed of a proprietary mix of non-ionic and zwitterionic detergents and Tris-based buffer. It allows disruption of E. coli cells without denaturing soluble proteins.
Catalog No. 50-217-2598
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.