Learn More
Ace Glass, Inc. DYNABLOC TUBE + VIAL + FLASK
SDP
Supplier:  Ace Glass, Inc. 1369611
Encompass_Preferred
Anodized aluminum Dynabloc heating/mixing blocks with three segments to hold up to three different size and type of vessels at the same time. Dynabloc has 1 (48.5mm diameter) well for 50mL round bottom flask, 10 (17x50mm) holes for 16x100mm tubes, and 4 (28x50mm) holes for 40mL vials. Dynablocs should be anchored to either a 135mm (50978108) or 145mm (50983541) base plate. A T-handle extractor (50978109) should be used to remove blocks when hot.
Catalog No. 50-978-104
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.