Learn More
Ace Glass, Inc. Dynabloc, heating block, flask, 250mL round bottom size, (3)62.2mm wells
Supplier: Ace Glass, Inc. 1369904
DynaBloc heating block with (3) 62.2mm wells, for use with 250mL round bottom flasks. Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/or heating of multiple samples contained within small, round bottom flasks. When hot, blocks should be removed using a T-handle DynaBloc extractor (Part 50978109).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.