Learn More
Ace Glass, Inc. Dynabloc, heating block, flask, 100mL round bottom size, (4)60.3mm wells
SDP
Supplier:  Ace Glass, Inc. 1369906
Encompass_Preferred
DynaBloc heating block with (4) 60.3mm wells, for use with 100mL round bottom flasks. Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/or heating of multiple samples contained within small, round bottom flasks. When hot, blocks should be removed using a T-handle DynaBloc extractor (Part 50978109).
Catalog No. 50-978-119
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.