Learn More
Ace Glass, Inc. Dynabloc block, 5mL V-vial size, 36 (22x35.6mm) holes
Supplier: Ace Glass, Inc. 1369837
5mL V-vial size Dynabloc block with 36 size 22x35.6mm holes. The Dynabloc heating system supplies low-cost, highly efficient, simultaneous stirring and/ or heating of multiple samples contained within various sizes of small vials and tubes.