Learn More
Proteintech Group Inc DYKDDDDK Fab-Trap™ Agarose
SDP
Supplier:  Proteintech Group Inc FFA100
Encompass
The ChromoTek DYKDDDDK Fab-Trap™ Agarose consists of an anti-DYKDDDDK-tag Fab-fragment, which is covalently bound to agarose beads. DYKDDDDK Fab-Trap™ Agarose is used to immunoprecipitate 1x and 3xDYKDDDDK-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc.
Catalog No. 50-225-8207
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.