Learn More
Electron Microscopy Sciences EMS Disposable Macro Knives Cartilage Premium, 4" Blade, 0.75" width
Supplier: Electron Microscopy Sciences 7205507
These disposable knives are economically priced and completely disposable. They have double honed, flexible blades with non-slip handles to ensure accurate dissection. They may be stropped for a longer lasting edge. They are available in standard or premium grades of Sheffield stainless steel.
Blade: 4"
Width: 0.75"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.