Learn More
Thermo Scientific™ DRI™ Phencyclidine (PCP) Drugs of Abuse Assays
Eliminate time-consuming reagent preparation yet deliver high analytical accuracy with Thermo Scientific™ DRI™ Phencyclidine (PCP) Drugs of Abuse Assays.
$1415.00 - $2667.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Multi-Drug Controls |
Detectable Analytes | Phencyclidine |
DoA Calibrators | DRI Multi-Drug Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-306
|
Thermo Scientific™
0160 |
1 x 100 mL |
Each for $2,667.00
|
|
|||||
23-278-353
|
Thermo Scientific™
10014673 |
3 x 18 mL |
Each for $1,415.00
|
|
|||||
Description
Eliminate time-consuming reagent preparation yet deliver high analytical accuracy with Thermo Scientific™ DRI™ Phencyclidine (PCP) Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Phencyclidine | |
Urine |
MGC Multi-Drug Controls | |
DRI Multi-Drug Calibrators | |
Reagent |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.