Learn More
Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays.
$2196.00 - $3166.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | DRI Oxycodone Control Set |
Detectable Analytes | Oxycodone |
DoA Calibrators | DRI Oxycodone Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-385
![]() |
Thermo Scientific™
100248 |
1 x 68 mL |
Each for $3,166.00
|
|
|||||
23-278-355
![]() |
Thermo Scientific™
10015632 |
3 x 18 mL |
Each for $2,196.00
|
|
|||||
Description
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. Supports new SAMHSA guidelines (effective October 1, 2017)
- Supports new SAMHSA guidelines (effective October 1, 2017)
- One of the largest menu of tests available
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
2°C to 8°C | |
Oxycodone | |
Urine |
DRI Oxycodone Control Set | |
DRI Oxycodone Calibrators | |
Reagent |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.