Learn More
Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators.
$180.00 - $187.00
Specifications
Description | DRI Methadone Metabolite Drugs of Abuse Calibrators |
---|---|
Detectable Analytes | EDDP |
Packaging | 1 Bottle |
Sample Type | Urine |
Content And Storage | 2°C to 8°C |
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
23-278-345
|
Thermo Scientific™
100117 |
150 ng/mL |
Each for $184.00
|
|
|||||
23-278-346
|
Thermo Scientific™
100118 |
300 ng/mL |
Each for $183.00
|
|
|||||
23-278-347
|
Thermo Scientific™
100120 |
1000 ng/mL |
Each for $187.00
|
|
|||||
23-278-348
|
Thermo Scientific™
100122 |
2000 ng/mL |
Each for $180.00
|
|
|||||
Description
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
DRI Methadone Metabolite Drugs of Abuse Calibrators | |
1 Bottle | |
2°C to 8°C | |
1 x 10 mL |
EDDP | |
Urine | |
Calibrator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.