Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Assay

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of DRI Methadone Metabolite Drugs of Abuse Assay.

$3303.00

Specifications

Content And Storage 2°C to 8°C
Sample Type Urine
Type Reagent
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-278-344
View Documents
Thermo Scientific™
100115
1 x 100 mL
Each of 1 for $3,303.00
Add to Cart
 
Description

Description

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
Specifications

Specifications

2°C to 8°C
Urine
Reagent
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.