Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators.

$180.00 - $184.00

Specifications

Detectable Analytes MDA, MDEA, MDMA
Packaging 1 Bottle
Sample Type Urine
Content And Storage 2°C to 8°C
Type Calibrator
View More Specs

Products 4
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
23-278-337
View Documents
Thermo Scientific™
100079
1000 ng/mL
Each for $183.00
missing translation for 'QuantityLeft'
Add to Cart
 
23-278-338
View Documents
Thermo Scientific™
100080
750 ng/mL
Each for $183.00
missing translation for 'QuantityLeft'
Add to Cart
 
23-278-466
View Documents
Thermo Scientific™
100081
500 ng/mL
Each for $180.00
missing translation for 'QuantityLeft'
Add to Cart
 
23-278-340
View Documents
Thermo Scientific™
100082
250 ng/mL
Each for $184.00
missing translation for 'QuantityLeft'
Add to Cart
 
Description

Description

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Ecstasy Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
Specifications

Specifications

MDA, MDEA, MDMA
Urine
Calibrator
1 Bottle
2°C to 8°C
1 x 10 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.