Learn More
Electron Microscopy Sciences DPX Mountant for Microscopy, 500ml
Supplier: Electron Microscopy Sciences 13512
DPX Mountant for Microscopy is a mixture of Distyrene, a plasticizer, and xylene. It is a colorless synthetic resin mounting media which replaces Xylene-Balsam. It preserves the stain and dries quickly. It comes in the amount of 500ml.