Learn More
Rat IgG (H+L) Highly Cross-Adsorbed Donkey anti-Rat, Alexa Fluor Plus 488, Secondary Antibody, Invitrogen™
Donkey Polyclonal Antibody
$162.00 - $362.00
Specifications
Antigen | Rat IgG (H+L) Highly Cross-Adsorbed |
---|---|
Concentration | 2 mg/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Alexa Fluor Plus 488 |
Description
To minimize cross-reactivity, the donkey anti-rat IgG whole antibodies have been cross-adsorbed against serum proteins from bovine, goat, rabbit, mouse, chicken, guinea pig, hamster, horse, sheep, and human to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment to eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.. This antibody binds to heavy chains on rat IgG and light chains on all rat immunoglobulins, does not bind non-immunoglobulin rat serum proteins or IgG from bovine, goat, rabbit, mouse, chicken, guinea pig, hamster, horse, sheep, or human.
Rat IgG (H+L) Highly Cross-Adsorbed Polyclonal antibody specifically detects Rat IgG (H+L) Highly Cross-Adsorbed in Rat samples. It is validated for Immunocytochemistry, ImmunofluorescenceSpecifications
Rat IgG (H+L) Highly Cross-Adsorbed | |
Immunocytochemistry | |
Alexa Fluor Plus 488 | |
Donkey | |
Rat | |
Gamma Immunoglobins Heavy and Light chains | |
Secondary | |
4° C, store in dark |
2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5 | |
IgG | |
Affinity chromatography |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.