Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dolastatin 10 trifluoroacetate

Tubulin polymerization inhibitor

Supplier:  Tocris Bioscience™ 3375/1

Catalog No. 33751


Explore all special offers
Add to Cart
This item is not returnable. View return policy

Description

Description

Dolastatin 10 trifluoroacetate is a tubulin polymerization inhibitor. Noncompetitively inhibits binding of radiolabeled vincristine to tubulin (Ki = 1.4 μM) and induces tubulin aggregation in vitro.
Specifications

Specifications

N,N-Dimethyl-L-valyl-N-[(1 S,2 R)-2-methoxy-4-[(2 S)-2-[(1 R,2 R)-1-methoxy-2-methyl-3-oxo-3-[[(1 S)-2-phenyl-1-(2-thiazol)ethyl]amino]propyl]-1-pyrrolidinyl]-1-[(1 S)-1-methylpropyl]-4-oxobutyl]-N-methyl--valinamide trifluoroacetate
1 mg
C42H68N6O6S.CF3CO2H
95%
2342568-65-2
Microtubule Binding Compounds
899.11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.