Learn More
Bethyl Laboratories, Inc DOG IGG2 AB, A40-121A 1MG
Supplier: Bethyl Laboratories, Inc A40121A
Sheep anti-Dog IgG2 Antibody Affinity Purified. Reactivity Dog.Applications WB, IHC, ICC, ELISAHost Sheep, Antibody Type Polyclonal. Unconjugated, Format Whole I.