Learn More
Chemglass Life Sciences DO Probe, w/VP, 12 x 325mm
SDP
Supplier:  Chemglass Life Sciences CLS1435127
Encompass_Preferred
DO Probe w/VP, 12x325mm. • Cathode: Platinum • Anode: Silver • Polarization: ~675mV • Output: 40 to 70nA in air at 25°C • Temperature: 0 to 60°C; Sterilization to 130°C • Stability: 2% or Readout per week at constant pressure/temp 30°C • Linearity: Better than ±0.3% of readout • Pressure: 60PSI maximum • Material: Wetted parts 316L SST • O-Ring: Viton
Catalog No. 50-143-147
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.