Learn More
Enzo Life Sciences AMPINEXT™ DNA Size Selection Kit (48Reactions)
SDP
Supplier:  Enzo Life Sciences ENZGEN5060048
Encompass_Preferred
The AMPINEXT™ DNA Size Selection Kit is a complete set of optimized reagents for quick removal of DNA fragments of <150 bps for library preparation in next generation sequencing applications. The kit utilizes magnetic bead technology and is suitable for Illumina, Life Technologies (SOLiD), Ion Torrent, and Roche/454 platforms. It can also be used for removing undesired larger DNA fragments by optimizing the bead to DNA volume ratio accordingly. The indicated number of reactions can be performed for a standard 50 µl solution input DNA sample. Assay Time: 30 minutes.
Catalog No. 50-201-3791
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.