Learn More
United States Biological Corporation DMEM LO GLUC W/O AMINOACIDPWD
SDP
Supplier:  United States Biological Corporation D98001325L
Encompass_Preferred
Dulbecco’s MEM is the most widely used modification of BME. It contains a 4-fold higher concentration of amino acids and vitamins. Non-essential amino acids and certain essential trace elements were added. The bicarbonate concentration was increased. The standard formula for DMEM is with 1000mg/ml glucose. DMEM was originally developed for the culture of mice embryonic cells. Today, it finds a broad application of serum free culture of normal and transformed mouse and chicken cells.
Catalog No. NC0697033
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.