Learn More
Enzo Life Sciences DL-threo-1-Phenyl-2-decanoylamino-3-morpholino-1-propanol (50 mg). CAS: 80938-69-8
Supplier: Enzo Life Sciences BMLSL2100050
Blocks glucosylation of ceramide by inhibiting UDP-glucose:ceramide glucosyltransferase (glucosylceramide synthetase). Inhibits the expression of cell surface glycolipid antigens and inhibits the growth of cultured rabbit skin fibroblasts. Possesses antitumor activity and inhibits Lewis lung carcinoma cell metastasis in vitro. Useful tool for studying the effects of cellular glycosphingolipid depletion. Enhances the apoptotic response to ionizing radiation by enhancing the ceramide signal. Alternative name: D,L-threo-PDMP . hydrochloride, DL-PDMP. Purity: ≥98% (TLC). Solubility: 25mg/ml soluble in 100% ethanol, DMSO or water (50°C, vortex or sonicate). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.