Learn More
Enzo Life Sciences DL-threo-1-Phenyl-2-palmitoylamino-3-morpholino-1-propanol (5 mg). CAS: 149022-18-4
Supplier: Enzo Life Sciences BMLSL2150005
Inhibits glucosylceramide synthetase. More effective than PDMP in intact cells. Alternative name: DL-PPMP. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: -20°C.