Learn More
Chemglass Life Sciences Distilling Adapter, Double, 10 Angled Side Neck, 45/50 Joint, 10mm PTFE Stopcock, 3/4" Swagelok
SDP
Supplier:  Chemglass Life Sciences CG123588
Encompass_Preferred
Distilling Adapter, Double, 10 Angled Side Neck, 45/50 Joint, 10mm PTFE Stopcock, 3/4" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10 angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 10mm PTFE stopcock with a 1/2 or 3/4 OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.
Catalog No. 50-153-1066
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.