Learn More
As One International Inc Diphtheria toxin, active, 200ug
Supplier:  As One International Inc 01517
Encompass_Preferred
Highly purified (>95% pure) from cultured medium of Corynebacterium diphtheriae strain PW8. It is a 53 kD protein consisting of two subunits linked by disulfide bridges It catalyzes ADP-ribosylation of eukaryotic elongation factor 2 and inhibits protein synthesis.Form:2-5 mg/ml in 20 mM Tris-HCl (pH 7.2) 150 mM NaCl10% GlycerolApplications:1. Inhibition of protein synthesis of eukaryotes 2. Putative anti-cancer drug utilyzing strong immunotoxin activity
Catalog No. 50-165-4924
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.