Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Dionex™ Fused Silica Tubing
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Supplier: Thermo Scientific™ 160481
Description
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Specifications
Fused Silica | |
LC-Packings Ultimate Capillary/Nano HPLC | |
Capillary Tubing | |
200 µm |
5 m | |
nanoLC | |
200 um I.D.; 280 um O.D.; 5 m length | |
280 µm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction